Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass Bottle
140-75-0
C7H8FN
125.15
IIFVWLUQBAIPMJ-UHFFFAOYSA-N
4-fluorobenzylamine,4-fluorophenyl methanamine,benzenemethanamine, 4-fluoro,p-fluorobenzylamine,benzylamine, p-fluoro,p-fluorobenzyl amine,4-fluoro-benzylamine,1-4-fluorophenyl methanamine,4-fluorobenzyl amine,chembl12392
67326
(4-fluorophenyl)methanamine
C1=CC(=CC=C1CN)F
140-75-0
C7H8FN
125.15
IIFVWLUQBAIPMJ-UHFFFAOYSA-N
4-fluorobenzylamine,4-fluorophenyl methanamine,benzenemethanamine, 4-fluoro,p-fluorobenzylamine,benzylamine, p-fluoro,p-fluorobenzyl amine,4-fluoro-benzylamine,1-4-fluorophenyl methanamine,4-fluorobenzyl amine,chembl12392
67326
(4-fluorophenyl)methanamine
C1=CC(=CC=C1CN)F
Infrared Spectrum Authentic
Refractive Index 1.5117
Density 1.0900g/mL
Boiling Point 183.0°C to 185.0°C
Quantity 500g
Specific Gravity 1.09
Packaging Glass Bottle
Flash Point 66°C
Color Colorless to Yellow
Assay Percent Range 96% min. (GC)
Linear Formula FC6H4CH2NH2
Formula Weight 125.15
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Fluorobenzylamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 205-430-4