Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Fluorobiphenyl-4-boronic acid, 98%

Click to view available options
Quantity:
250mg
1g
5g
140369-67-1
C12H10BFO2
216.018
MFCD08544387
KHMFYFVXTICBEL-UHFFFAOYSA-N
4-4-fluorophenyl phenylboronic acid,4'-fluoro-1,1'-biphenyl-4-yl boronic acid,4'-fluoro-4-biphenylboronic acid,4'-fluorobiphenyl-4-ylboronic acid,4-4-fluorophenyl phenylboronicacid,4'-fluorobiphenyl-4-boronic acid,boronic acid,b-4'-fluoro 1,1'-biphenyl-4-yl,4'-fluoro-1,1'-biphenyl-4-ylboronic acid,akos brn-1038,pubchem11575
14985212
[4-(4-fluorophenyl)phenyl]boronic acid
B(C1=CC=C(C=C1)C2=CC=C(C=C2)F)(O)O
140369-67-1
C12H10BFO2
216.018
MFCD08544387
KHMFYFVXTICBEL-UHFFFAOYSA-N
4-4-fluorophenyl phenylboronic acid,4'-fluoro-1,1'-biphenyl-4-yl boronic acid,4'-fluoro-4-biphenylboronic acid,4'-fluorobiphenyl-4-ylboronic acid,4-4-fluorophenyl phenylboronicacid,4'-fluorobiphenyl-4-boronic acid,boronic acid,b-4'-fluoro 1,1'-biphenyl-4-yl,4'-fluoro-1,1'-biphenyl-4-ylboronic acid,akos brn-1038,pubchem11575
14985212
[4-(4-fluorophenyl)phenyl]boronic acid
B(C1=CC=C(C=C1)C2=CC=C(C=C2)F)(O)O
Quantity 250mg
Beilstein 8689987
Formula Weight 216.02
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4'-Fluorobiphenyl-4-boronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C