Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Fluorobiphenyl-4-sulfonyl chloride, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
116748-66-4
C12H8ClFO2S
270.71
CIDMHDJTWVMBIF-UHFFFAOYSA-N
4'-fluorobiphenyl-4-sulfonyl chloride,4-4-fluorophenyl benzenesulfonyl chloride,4'-fluoro 1,1'-biphenyl-4-sulfonyl chloride,4'-fluoro-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride, 4'-fluoro,4'-fluoro-1,1'-biphenyl-4-sulfonyl chloride,acmc-20apun,4'-fluorobiphenyl-4-sulfonylchloride,4'-fluoro-4-biphenylsulfonyl chloride
2794749
4-(4-fluorophenyl)benzenesulfonyl chloride
C1=CC(=CC=C1C2=CC=C(C=C2)S(=O)(=O)Cl)F
116748-66-4
C12H8ClFO2S
270.71
CIDMHDJTWVMBIF-UHFFFAOYSA-N
4'-fluorobiphenyl-4-sulfonyl chloride,4-4-fluorophenyl benzenesulfonyl chloride,4'-fluoro 1,1'-biphenyl-4-sulfonyl chloride,4'-fluoro-biphenyl-4-sulfonyl chloride,1,1'-biphenyl-4-sulfonylchloride, 4'-fluoro,4'-fluoro-1,1'-biphenyl-4-sulfonyl chloride,acmc-20apun,4'-fluorobiphenyl-4-sulfonylchloride,4'-fluoro-4-biphenylsulfonyl chloride
2794749
4-(4-fluorophenyl)benzenesulfonyl chloride
C1=CC(=CC=C1C2=CC=C(C=C2)S(=O)(=O)Cl)F
Quantity 5g
Melting Point 82.0°C to 84.0°C
Assay Percent Range 96% min. (GC)
Formula Weight 270.71
Percent Purity 97%
Chemical Name or Material 4'-Fluorobiphenyl-4-sulfonyl chloride

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger