Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoroisoindoline hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
924305-06-6
C8H9ClFN
173.615
MKNPYNGKUKUSFB-UHFFFAOYSA-N
4-fluoroisoindoline hydrochloride,4-fluoro-2,3-dihydro-1h-isoindole hydrochloride,4-fluoro-isoindoline hcl,4-fluoro-1h-isoindoline hydrochloride,4-fluoroisoindolinehydrochloride,4-fluoro isoindoline hydrochloride,4-fluoro-isoindoline hydrochloride,fluoroisoindoline-hydrochloride salt
42609643
4-fluoro-2,3-dihydro-1H-isoindole;hydrochloride
C1C2=C(CN1)C(=CC=C2)F.Cl
924305-06-6
C8H9ClFN
173.615
MKNPYNGKUKUSFB-UHFFFAOYSA-N
4-fluoroisoindoline hydrochloride,4-fluoro-2,3-dihydro-1h-isoindole hydrochloride,4-fluoro-isoindoline hcl,4-fluoro-1h-isoindoline hydrochloride,4-fluoroisoindolinehydrochloride,4-fluoro isoindoline hydrochloride,4-fluoro-isoindoline hydrochloride,fluoroisoindoline-hydrochloride salt
42609643
4-fluoro-2,3-dihydro-1H-isoindole;hydrochloride
C1C2=C(CN1)C(=CC=C2)F.Cl
Water 5% Max. (K.F.)
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 173.62
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-Fluoroisoindoline hydrochloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.