Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluoronaphthalene-1-carbonitrile, 97%

Click to view available options
Quantity:
1g
5g
25g
13916-99-9
C11H6FN
171.174
MFCD00236656
XQSGYYNFKIQWAC-UHFFFAOYSA-N
1-cyano-4-fluoronaphthalene,4-fluoro-1-naphthonitrile,4-fluoro-1-naphthalenecarbonitrile,1-naphthalenecarbonitrile,4-fluoro,1-naphthalenecarbonitrile, 4-fluoro,4-fluoronaphthalenecarbonitrile,4-fluoro-1-phthonitrile,acmc-1buk6,1-cyano-4-fluoronapthalene,4-cyano-1-fluoronaphthalene
2736734
4-fluoronaphthalene-1-carbonitrile
C1=CC=C2C(=C1)C(=CC=C2F)C#N
13916-99-9
C11H6FN
171.174
MFCD00236656
XQSGYYNFKIQWAC-UHFFFAOYSA-N
1-cyano-4-fluoronaphthalene,4-fluoro-1-naphthonitrile,4-fluoro-1-naphthalenecarbonitrile,1-naphthalenecarbonitrile,4-fluoro,1-naphthalenecarbonitrile, 4-fluoro,4-fluoronaphthalenecarbonitrile,4-fluoro-1-phthonitrile,acmc-1buk6,1-cyano-4-fluoronapthalene,4-cyano-1-fluoronaphthalene
2736734
4-fluoronaphthalene-1-carbonitrile
C1=CC=C2C(=C1)C(=CC=C2F)C#N
Melting Point 89°C to 91°C
Quantity 5g
UN Number UN3439
Formula Weight 171.18
Percent Purity 97%
Chemical Name or Material 4-Fluoronaphthalene-1-carbonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.