Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenol 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
371-41-5
C6H5FO
112.1
MFCD00002316
RHMPLDJJXGPMEX-UHFFFAOYSA-N
p-fluorophenol,phenol, 4-fluoro,phenol, p-fluoro,para-fluorophenol,4-fluoro phenol,4-fluoro-phenol,ccris 665,4-fluoranylphenol,fpn
9732
4-fluorophenol
C1=CC(=CC=C1O)F
371-41-5
C6H5FO
112.1
MFCD00002316
RHMPLDJJXGPMEX-UHFFFAOYSA-N
p-fluorophenol,phenol, 4-fluoro,phenol, p-fluoro,para-fluorophenol,4-fluoro phenol,4-fluoro-phenol,ccris 665,4-fluoranylphenol,fpn
9732
4-fluorophenol
C1=CC(=CC=C1O)F
Infrared Spectrum Authentic
Color Beige-Yellow to Pink
Boiling Point 185.0°C
Packaging Glass bottle
Flash Point 68°C
Melting Point 44.0°C to 49.0°C
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula FC6H4OH
Beilstein 06, 183
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in methylene dichloride
Formula Weight 112.1
Physical Form Crystalline Solid
Percent Purity 99%
Chemical Name or Material 4-Fluorophenol, 99%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-736-0

RTECSNumber : SL4550000

TSCA : TSCA