Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenoxy-ethylbromide, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
332-48-9
C8H8BrFO
219.05
MFCD00044739
JXSPKRUNMHMICQ-UHFFFAOYSA-N
1-2-bromoethoxy-4-fluorobenzene,4-fluorophenoxy-ethylbromide,2-bromoethyl 4-fluorophenyl ether,4-fluorophenoxyethylbromide,4-2-bromoethoxy fluorobenzene,4-fluorophenoxyethyl bromide,beta-bromo-4-fluorophenetole,2-4-fluorophenoxy ethyl bromide,benzene, 1-2-bromoethoxy-4-fluoro,p-fluorophenoxy ethylbromide
2064171
1-(2-bromoethoxy)-4-fluorobenzene
C1=CC(=CC=C1OCCBr)F
332-48-9
C8H8BrFO
219.05
MFCD00044739
JXSPKRUNMHMICQ-UHFFFAOYSA-N
1-2-bromoethoxy-4-fluorobenzene,4-fluorophenoxy-ethylbromide,2-bromoethyl 4-fluorophenyl ether,4-fluorophenoxyethylbromide,4-2-bromoethoxy fluorobenzene,4-fluorophenoxyethyl bromide,beta-bromo-4-fluorophenetole,2-4-fluorophenoxy ethyl bromide,benzene, 1-2-bromoethoxy-4-fluoro,p-fluorophenoxy ethylbromide
2064171
1-(2-bromoethoxy)-4-fluorobenzene
C1=CC(=CC=C1OCCBr)F
Refractive Index 1.5300 to 1.532
Packaging Glass bottle
Color Brown to Colorless
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula FC6H4O(CH2)2Br
Formula Weight 219.05
Physical Form Liquid After Melting
Percent Purity ≥94%
Chemical Name or Material 4-Fluorophenoxy-ethylbromide, 95%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.