Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenyl isocyanate, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
1195-45-5
C7H4FNO
137.11
MFCD00002023
DSVGFKBFFICWLZ-UHFFFAOYSA-N
4-fluorophenyl isocyanate,4-fluorophenylisocyanate,p-fluorophenyl isocyanate,benzene, 1-fluoro-4-isocyanato,isocyanic acid 4-fluorophenyl ester,1-fluoro-4-isocyanato-benzene,benzene,1-fluoro-4-isocyanato,4-fluorobenzenisocyanate,paragos 331046,timtec-bb sbb032971
70955
1-fluoro-4-isocyanatobenzene
C1=CC(=CC=C1N=C=O)F
1195-45-5
C7H4FNO
137.11
MFCD00002023
DSVGFKBFFICWLZ-UHFFFAOYSA-N
4-fluorophenyl isocyanate,4-fluorophenylisocyanate,p-fluorophenyl isocyanate,benzene, 1-fluoro-4-isocyanato,isocyanic acid 4-fluorophenyl ester,1-fluoro-4-isocyanato-benzene,benzene,1-fluoro-4-isocyanato,4-fluorobenzenisocyanate,paragos 331046,timtec-bb sbb032971
70955
1-fluoro-4-isocyanatobenzene
C1=CC(=CC=C1N=C=O)F
Boiling Point 68°C
Specific Gravity 1.206
Flash Point 60°C
Refractive Index 1.5141 to 1.5161
Density 1.2060g/mL
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 25mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula FC6H4NCO
Solubility Information Solubility in water: decomposes
Formula Weight 137.11
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 4-Fluorophenyl isocyanate, 99%
Show More Show Less

GHS H Statement:
Fatal if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-799-0