Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenylacetic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
405-50-5
C8H7FO2
154.14
MFCD00004343
MGKPFALCNDRSQD-UHFFFAOYSA-N
4-fluorophenylacetic acid,2-4-fluorophenyl acetic acid,p-fluorophenylacetic acid,4-fluorophenyl acetic acid,benzeneacetic acid, 4-fluoro,4-fluorobenzeneacetic acid,p-fluorophenyl acetic acid,acetic acid, p-fluorophenyl,unii-s5yjv7caz1,4-fluoro-phenyl-acetic acid
9837
2-(4-fluorophenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)F
405-50-5
C8H7FO2
154.14
MFCD00004343
MGKPFALCNDRSQD-UHFFFAOYSA-N
4-fluorophenylacetic acid,2-4-fluorophenyl acetic acid,p-fluorophenylacetic acid,4-fluorophenyl acetic acid,benzeneacetic acid, 4-fluoro,4-fluorobenzeneacetic acid,p-fluorophenyl acetic acid,acetic acid, p-fluorophenyl,unii-s5yjv7caz1,4-fluoro-phenyl-acetic acid
9837
2-(4-fluorophenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)F
Quantity 25g
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Melting Point 85.0°C to 86.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula FC6H4CH2CO2H
Merck Index 15, 4209
Formula Weight 154.14
Physical Form Shiny Crystalline Powder or Flakes
Percent Purity 98%
Chemical Name or Material 4-Fluorophenylacetic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.