Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenylacetonitrile 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
459-22-3
C8H6FN
135.14
MFCD00001917
JHQBLYITVCBGTO-UHFFFAOYSA-N
4-fluorophenylacetonitrile,4-fluorobenzyl cyanide,p-fluorobenzyl cyanide,benzeneacetonitrile, 4-fluoro,4-fluorobenzylcyanide,4-fluorobenzeneacetonitrile,p-fluorophenylacetonitrile,2-4-fluorophenyl acetonitrile,4-fluorobenzylnitrile,acetonitrile, p-fluorophenyl
68016
2-(4-fluorophenyl)acetonitrile
C1=CC(=CC=C1CC#N)F
459-22-3
C8H6FN
135.14
MFCD00001917
JHQBLYITVCBGTO-UHFFFAOYSA-N
4-fluorophenylacetonitrile,4-fluorobenzyl cyanide,p-fluorobenzyl cyanide,benzeneacetonitrile, 4-fluoro,4-fluorobenzylcyanide,4-fluorobenzeneacetonitrile,p-fluorophenylacetonitrile,2-4-fluorophenyl acetonitrile,4-fluorobenzylnitrile,acetonitrile, p-fluorophenyl
68016
2-(4-fluorophenyl)acetonitrile
C1=CC(=CC=C1CC#N)F
Flash Point 108°C
Boiling Point 119°C to 120°C (18.0mmHg)
Specific Gravity 1.126
Refractive Index 1.4990 to 1.502
Density 1.1260g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 100g
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula FC6H4CH2CN
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene and dichloromethane
Formula Weight 135.14
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 4-Fluorophenylacetonitrile, 95%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-286-8

RTECSNumber : AM0210000

TSCA : TSCA