Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenylacetyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
459-04-1
C8H6ClFO
172.59
MFCD00674181
SIOJFYRPBYGHOO-UHFFFAOYSA-N
4-fluorophenylacetyl chloride,2-4-fluorophenyl acetyl chloride,4-fluorophenylacetylchloride,4-fluorobenzeneacetyl chloride,p-fluorophenylacetyl chloride,benzeneacetyl chloride, 4-fluoro,2-4-fluorophenyl ethanoyl chloride,4-fluoro phenylacetyl chloride,pubchem17517,acmc-1aivq
3744988
2-(4-fluorophenyl)acetyl chloride
C1=CC(=CC=C1CC(=O)Cl)F
459-04-1
C8H6ClFO
172.59
MFCD00674181
SIOJFYRPBYGHOO-UHFFFAOYSA-N
4-fluorophenylacetyl chloride,2-4-fluorophenyl acetyl chloride,4-fluorophenylacetylchloride,4-fluorobenzeneacetyl chloride,p-fluorophenylacetyl chloride,benzeneacetyl chloride, 4-fluoro,2-4-fluorophenyl ethanoyl chloride,4-fluoro phenylacetyl chloride,pubchem17517,acmc-1aivq
3744988
2-(4-fluorophenyl)acetyl chloride
C1=CC(=CC=C1CC(=O)Cl)F
Refractive Index 1.5110 to 1.513
Density 1.2590g/mL
Boiling Point 60°C (8.0mmHg)
Flash Point 107°C
Specific Gravity 1.259
Infrared Spectrum Authentic
Color Yellow
Quantity 25g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula FC6H4CH2C(O)Cl
Solubility Information Solubility in water: reacts
Formula Weight 172.59
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Fluorophenylacetyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger