Alfa Aesar™ (4-Fluorophenylethynyl)trimethylsilane, 97%

Manufacturer: Alfa Aesar™ H55688AA

Quantity 5mL

 View more versions of this product

Catalog No. AAH55688AA

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 130995-12-9
    Formula Weight 192.31
    MDL Number MFCD03701552
    Chemical Name or Material (4-Fluorophenylethynyl)trimethylsilane

    Specifications

    Formula Weight 192.31
    MDL Number MFCD03701552
    Physical Form Liquid
    Refractive Index 1.5090
    CAS 130995-12-9
    Color Colorless
    Boiling Point 40°C (0.1mmHg)
    Quantity 5mL
    Density 0.948g/mL
    Chemical Name or Material (4-Fluorophenylethynyl)trimethylsilane
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit