Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenylhydrazine hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
823-85-8
C6H7FN2·HCl
162.59
MFCD00012942
FEKUXLUOKFSMRO-UHFFFAOYSA-N
4-fluorophenylhydrazine hydrochloride,4-fluorophenyl hydrazine hydrochloride,4-fluorophenylhydrazine hcl,1-4-fluorophenyl hydrazine hydrochloride,4-fluorophenyl hydrazine hcl,p-fluorophenylhydrazine hydrochloride,4-fluorphenyl hydrazine hydrochloride,4-fluorophenyl hydrazine monohydrochloride,p-fluorophenylhydrazine hcl,hydrazine, 4-fluorophenyl-, monohydrochloride
69981
(4-fluorophenyl)hydrazine;hydrochloride
C1=CC(=CC=C1NN)F.Cl
823-85-8
C6H7FN2·HCl
162.59
MFCD00012942
FEKUXLUOKFSMRO-UHFFFAOYSA-N
4-fluorophenylhydrazine hydrochloride,4-fluorophenyl hydrazine hydrochloride,4-fluorophenylhydrazine hcl,1-4-fluorophenyl hydrazine hydrochloride,4-fluorophenyl hydrazine hcl,p-fluorophenylhydrazine hydrochloride,4-fluorphenyl hydrazine hydrochloride,4-fluorophenyl hydrazine monohydrochloride,p-fluorophenylhydrazine hcl,hydrazine, 4-fluorophenyl-, monohydrochloride
69981
(4-fluorophenyl)hydrazine;hydrochloride
C1=CC(=CC=C1NN)F.Cl
Melting Point 275°C to 277°C
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Brown to White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula FC6H4NHNH2·HCl
Beilstein 15, I, 105
Solubility Information Solubility in water: soluble
Formula Weight 162.59
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-Fluorophenylhydrazine hydrochloride
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Harmful if inhaled.
Suspected of causing cancer.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning