Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenylmagnesium bromide, 0.8M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
352-13-6,109-99-9
C6H4BrFMg
199.305
MFCD00009667
BRKADVNLTRCLOW-UHFFFAOYSA-M
4-fluorophenylmagnesium bromide,magnesium, bromo 4-fluorophenyl,bromo 4-fluorophenyl magnesium,p-fluorophenylmagnesium bromide,4-fluorophenyl magnesium bromide,4-f-c6h4-mgbr,4-fluorophenylmagnesiumbromide,4-fluorphenylmagnesium bromide,4-flourophenylmagnesium bromide,4-fluorophenyl magnesiumbromide
2734897
magnesium;fluorobenzene;bromide
C1=CC(=CC=[C-]1)F.[Mg+2].[Br-]
352-13-6,109-99-9
C6H4BrFMg
199.305
MFCD00009667
BRKADVNLTRCLOW-UHFFFAOYSA-M
4-fluorophenylmagnesium bromide,magnesium, bromo 4-fluorophenyl,bromo 4-fluorophenyl magnesium,p-fluorophenylmagnesium bromide,4-fluorophenyl magnesium bromide,4-f-c6h4-mgbr,4-fluorophenylmagnesiumbromide,4-fluorphenylmagnesium bromide,4-flourophenylmagnesium bromide,4-fluorophenyl magnesiumbromide
2734897
magnesium;fluorobenzene;bromide
C1=CC(=CC=[C-]1)F.[Mg+2].[Br-]
Linear Formula F(C6H6)MgBr
Density 1.0100g/mL
Formula Weight 199.3
Physical Form Solution
Percent Purity 14 to 17%
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: vigorous reaction.
Specific Gravity 1.01
Storage Information product may develop some turbidity or precipitate during storage
Color Yellow to Brown
Melting Point 5.0°C
Quantity 100mL
Flash Point −21°C
Chemical Name or Material 4-Fluorophenylmagnesium bromide, 0.8M Solution in THF
Show More Show Less

GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Highly flammable liquid and vapor. In contact with water releases flammable gas. Reacts violently with water. May form explosive peroxides.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger