Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluorophenylmagnesium bromide, 1M in MeTHF

Manufacturer:  Alfa Aesar™ H51169AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH51169AP


  Check Availability
Add to cart

Specifications

Description

Applications
4-Fluorophenylmagnesium bromide is used as a reagent for the introduction of the 4-Fluorophenyl group.

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

352-13-6
199.31
UN2924
C6H4BrFMg
MFCD00009667
4-Fluorophenylmagnesium bromide

Specifications

1M in MeTHF
352-13-6
199.31
UN2924
500mL
C6H4BrFMg
MFCD00009667
4-Fluorophenylmagnesium bromide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit