Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluorophthalonitrile, 98%

Click to view available options
Quantity:
250mg
1g
5g
65610-14-2
C8H3FN2
146.124
MFCD00210430
QQEKYCCJLSRLEC-UHFFFAOYSA-N
4-fluorophthalonitrile,1,2-benzenedicarbonitrile, 4-fluoro,4-fluorophthalodinitrile,1,2-dicyano-4-fluorobenzene,4-fluoro-phthalonitrile,2-cyano-5-fluorobenzonitrile,4-fluorophthalonitrle,pubchem4027,acmc-1bb7s
103423
4-fluorobenzene-1,2-dicarbonitrile
C1=CC(=C(C=C1F)C#N)C#N
65610-14-2
C8H3FN2
146.124
MFCD00210430
QQEKYCCJLSRLEC-UHFFFAOYSA-N
4-fluorophthalonitrile,1,2-benzenedicarbonitrile, 4-fluoro,4-fluorophthalodinitrile,1,2-dicyano-4-fluorobenzene,4-fluoro-phthalonitrile,2-cyano-5-fluorobenzonitrile,4-fluorophthalonitrle,pubchem4027,acmc-1bb7s
103423
4-fluorobenzene-1,2-dicarbonitrile
C1=CC(=C(C=C1F)C#N)C#N
Quantity 250mg
Melting Point 100°C to 102°C
UN Number UN3439
Beilstein 2443236
Formula Weight 146.12
Percent Purity 98%
Chemical Name or Material 4-Fluorophthalonitrile

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.