Learn More
Aat BioquestSupplier Diversity Partner 4-Fluororesorcinol *CAS#: 103068-41-3*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 4022
4-Fluororesorcinol is a precursor that can be used for preparing fluorescent dyes such as fluorescein, coumarin and resorufin analogs. It is the key precusor for preparing Oregon Green dyes.
Catalog No. 50-195-3232