Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Fluorosalicylic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
345-29-9
C7H5FO3
156.112
MFCD00153168
TTZOLDXHOCCNMF-UHFFFAOYSA-N
4-fluorosalicylic acid,4-fluorosalicyclic acid,2-hydroxy-4-fluorobenzoic acid,4-fluoro-2-hydroxy-benzoic acid,fluorohydroxybenzoicacid5,benzoic acid, 4-fluoro-2-hydroxy,4-fluorosalicylicacid,zlchem 109,pubchem3493
67661
4-fluoro-2-hydroxybenzoic acid
C1=CC(=C(C=C1F)O)C(=O)O
345-29-9
C7H5FO3
156.112
MFCD00153168
TTZOLDXHOCCNMF-UHFFFAOYSA-N
4-fluorosalicylic acid,4-fluorosalicyclic acid,2-hydroxy-4-fluorobenzoic acid,4-fluoro-2-hydroxy-benzoic acid,fluorohydroxybenzoicacid5,benzoic acid, 4-fluoro-2-hydroxy,4-fluorosalicylicacid,zlchem 109,pubchem3493
67661
4-fluoro-2-hydroxybenzoic acid
C1=CC(=C(C=C1F)O)C(=O)O
Applications
4-Fluorosalicylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Quantity 5g
Melting Point 186°C to 190°C
Beilstein 2209124
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 4-Fluorosalicylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning