Alfa Aesar™ (+/-)-4-Fluorostyrene oxide, 98%

Manufacturer: Alfa Aesar™ H2660006

Quantity 5g

 View more versions of this product

Catalog No. AAH2660006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18511-62-1
    Molecular Formula C8H7FO
    Formula Weight 138.14
    MDL Number MFCD00043994
    UN Number UN2810
    Synonym (+/-)-2-(4-Fluorophenyl)oxirane
    Chemical Name or Material (+/-)-4-Fluorostyrene oxide

    Specifications

    Formula Weight 138.14
    Molecular Formula C8H7FO
    MDL Number MFCD00043994
    CAS 18511-62-1
    UN Number UN2810
    Synonym (+/-)-2-(4-Fluorophenyl)oxirane
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H351-H312-H332-H319-H227
    Suspected of causing cancer.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Flash Point 77°C (171°F)
    Odor Pungent
    Boiling Point 92°C (14mmHg)
    Quantity 5g
    Chemical Name or Material (+/-)-4-Fluorostyrene oxide
    Assay 98%

    Description

    Applications
    (+/-)-4-Fluorostyrene oxide used in chemicals, pharmaceutical intermediates. It is also used in organic chemistry that has also many industrial applications, biotransformation

    Solubility
    Difficult to mix.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H351-H312-H332-H319-H227
    Suspected of causing cancer.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes serious eye irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit