Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-4-Fluorostyrene oxide, 98%

Manufacturer:  Alfa Aesar™ H2660014

Quantity 25g

 View more versions of this product

Catalog No. AAH2660014


  Check Availability
Add to cart

Specifications

Description

Applications
(+/-)-4-Fluorostyrene oxide used in chemicals, pharmaceutical intermediates. It is also used in organic chemistry that has also many industrial applications, biotransformation

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

18511-62-1
138.14
UN2810
(+/-)-4-Fluorostyrene oxide
C8H7FO
MFCD00043994
(+/-)-2-(4-Fluorophenyl)oxirane

Specifications

77°C (171°F)
92°C (14mmHg)
18511-62-1
138.14
UN2810
(+/-)-4-Fluorostyrene oxide
Pungent
25g
C8H7FO
MFCD00043994
(+/-)-2-(4-Fluorophenyl)oxirane
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H351-H312-H332-H319-H227
Suspected of causing cancer.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit