Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-fluorothioanisole 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
371-15-3
C7H7FS
142.191
MFCD00040829
XFUMHENRNCUHOH-UHFFFAOYSA-N
4-fluorothioanisole,1-fluoro-4-methylthio benzene,4-fluorophenyl methyl sulfide,4-fluoro thioanisole,1-fluoro-4-methylsulfanyl-benzene,1-fluoro-4-methylsulfanyl benzene,p-fluoro methylthio benzene,benzene, 1-fluoro-4-methylthio,4-fluoro-1-methylthiobenzene,pubchem10641
3083565
1-fluoro-4-methylsulfanylbenzene
CSC1=CC=C(C=C1)F
371-15-3
C7H7FS
142.191
MFCD00040829
XFUMHENRNCUHOH-UHFFFAOYSA-N
4-fluorothioanisole,1-fluoro-4-methylthio benzene,4-fluorophenyl methyl sulfide,4-fluoro thioanisole,1-fluoro-4-methylsulfanyl-benzene,1-fluoro-4-methylsulfanyl benzene,p-fluoro methylthio benzene,benzene, 1-fluoro-4-methylthio,4-fluoro-1-methylthiobenzene,pubchem10641
3083565
1-fluoro-4-methylsulfanylbenzene
CSC1=CC=C(C=C1)F
Refractive Index 1.5500 to 1.5520 (20°C, 589nm)
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
Density 1.1670g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula FC6H4SCH3
Formula Weight 142.2
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 4-Fluorothioanisole
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning