Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Fluorovalerophenone, 97%

Click to view available options
Quantity:
5g
29114-66-7
C11H13FO
MFCD00017968
n-Butyl 4-fluorophenyl ketone
29114-66-7
C11H13FO
MFCD00017968
n-Butyl 4-fluorophenyl ketone
Melting Point 25°C to 28°C
Quantity 5g
Boiling Point 119°C to 123°C (22mmHg)
Beilstein 2502250
Formula Weight 180.21
Percent Purity 97%
Chemical Name or Material 4'-Fluorovalerophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C