Alfa Aesar™ 4-(Fmoc-amino)benzoic acid, 97%

Manufacturer: Alfa Aesar™ H5282806

Quantity 5g

 View more versions of this product

Catalog No. AAH5282806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 185116-43-2
    Molecular Formula C22H17NO4
    Formula Weight 359.38
    MDL Number MFCD00144888
    Beilstein 7722986
    Synonym Fmoc-4-Abz-OH
    Chemical Name or Material 4-(Fmoc-amino)benzoic acid

    Specifications

    Formula Weight 359.38
    Molecular Formula C22H17NO4
    MDL Number MFCD00144888
    CAS 185116-43-2
    Beilstein 7722986
    Synonym Fmoc-4-Abz-OH
    Quantity 5g
    Chemical Name or Material 4-(Fmoc-amino)benzoic acid
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit