Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Fmoc-aminomethyl)benzoic acid, 95%

Click to view available options
Quantity:
5g
25g
164470-64-8
C23H19NO4
373.408
MFCD01074693
JRHUROPSUJVMNH-UHFFFAOYSA-N
fmoc-4-amb-oh,4-9h-fluoren-9-yl methoxy carbonyl amino methyl benzoic acid,4-9h-fluoren-9-ylmethoxycarbonylamino methyl benzoic acid,4-9h-fluoren-9-ylmethoxy carbonyl amino methyl benzoic acid,4-9h-fluoren-9-ylmethoxy carbonyl amino methyl benzoic acid,4-9h-fluoren-9-yl methoxy carbonyl amino methyl benzoic acid,4-n-fmoc-aminomethyl-benzoic acid,4-fmoc-aminomethyl benzoicacid,4-fmoc-aminomethyl benzoic acid,fmoc-4-amb-oh fmoc-4-aminomethyl benzoic acid
2756083
4-[(9H-fluoren-9-ylmethoxycarbonylamino)methyl]benzoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCC4=CC=C(C=C4)C(=O)O
164470-64-8
C23H19NO4
373.408
MFCD01074693
JRHUROPSUJVMNH-UHFFFAOYSA-N
fmoc-4-amb-oh,4-9h-fluoren-9-yl methoxy carbonyl amino methyl benzoic acid,4-9h-fluoren-9-ylmethoxycarbonylamino methyl benzoic acid,4-9h-fluoren-9-ylmethoxy carbonyl amino methyl benzoic acid,4-9h-fluoren-9-ylmethoxy carbonyl amino methyl benzoic acid,4-9h-fluoren-9-yl methoxy carbonyl amino methyl benzoic acid,4-n-fmoc-aminomethyl-benzoic acid,4-fmoc-aminomethyl benzoicacid,4-fmoc-aminomethyl benzoic acid,fmoc-4-amb-oh fmoc-4-aminomethyl benzoic acid
2756083
4-[(9H-fluoren-9-ylmethoxycarbonylamino)methyl]benzoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCC4=CC=C(C=C4)C(=O)O
Applications
4-(Fmoc-aminomethyl)benzoic acid is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Incompatible with heat and oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Quantity 25g
Color White
Melting Point 224°C to 228°C
Beilstein 7391207
Formula Weight 373.41
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 4-(Fmoc-aminomethyl)benzoic acid

storageNote1 : 2° to 8°C