Learn More
4-Formylmorpholine, 99%, Thermo Scientific Chemicals
Description
4-Formylmorpholine is widely used as a solvent and extracting agent for aromatics. It is used as a scrubbing agent for acidic gases. It is employed in the preparation of adenine hydrochloride labeled with 14C, 4-(trifluoromethyl)morpholine, 3-alkyl-2-chloro-5,6-dihydrobenzaldehyde and terphenyl dialdehyde. It plays an important role in the desulfurization of natural gas, synthetic gas, flue gas and gasoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Formylmorpholine is widely used as a solvent and extracting agent for aromatics. It is used as a scrubbing agent for acidic gases. It is employed in the preparation of adenine hydrochloride labeled with 14C, 4-(trifluoromethyl)morpholine, 3-alkyl-2-chloro-5,6-dihydrobenzaldehyde and terphenyl dialdehyde. It plays an important role in the desulfurization of natural gas, synthetic gas, flue gas and gasoline.
Solubility
Miscible with water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 20°C to 23°C |
Density | 1.15 |
pH | 10 |
Boiling Point | 239°C to 241°C |
Flash Point | 118°C (244°F) |
Refractive Index | 1.486 |
Quantity | 2500 g |
Beilstein | 110293 |
Sensitivity | Hygroscopic |
Solubility Information | Miscible with water. |
Show More |
Safety and Handling
Hazard Category | H315-H320 |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P501c |
EINECSNumber | 224-518-3 |
RTECSNumber | QD9694000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.