Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Formylmorpholine, 99%

Click to view available options
Quantity:
100g
500g
2500g
4394-85-8
C5H9NO2
115.132
MFCD00006170
LCEDQNDDFOCWGG-UHFFFAOYSA-N
4-formylmorpholine,n-formylmorpholine,4-morpholinecarboxaldehyde,morpholine, 4-formyl,n-formylmorfolin,4-morpholinecarbaldehyde,n-formylmorfolin czech,unii-d1e63xo4rh,n-formyhnorpholine,n-formyl morpholin
20417
CHEBI:43989
morpholine-4-carbaldehyde
C1COCCN1C=O
4394-85-8
C5H9NO2
115.132
MFCD00006170
LCEDQNDDFOCWGG-UHFFFAOYSA-N
4-formylmorpholine,n-formylmorpholine,4-morpholinecarboxaldehyde,morpholine, 4-formyl,n-formylmorfolin,4-morpholinecarbaldehyde,n-formylmorfolin czech,unii-d1e63xo4rh,n-formyhnorpholine,n-formyl morpholin
20417
CHEBI:43989
morpholine-4-carbaldehyde
C1COCCN1C=O
Applications
4-Formylmorpholine is widely used as a solvent and extracting agent for aromatics. It is used as a scrubbing agent for acidic gases. It is employed in the preparation of adenine hydrochloride labeled with 14C, 4-(trifluoromethyl)morpholine, 3-alkyl-2-chloro-5,6-dihydrobenzaldehyde and terphenyl dialdehyde. It plays an important role in the desulfurization of natural gas, synthetic gas, flue gas and gasoline.

Solubility
Miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Boiling Point 239°C to 241°C
Melting Point 20°C to 23°C
Flash Point 118°C (244°F)
Quantity 500g
pH 10
Beilstein 110293
Formula Weight 115.13
Percent Purity 99%
Chemical Name or Material 4-Formylmorpholine

RTECSNumber : QD9694000

TSCA : TSCA