Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ 4 ft. Cell Logic™ Class II, Type A2 Biosafety Cabinet, US Model
SDP

Catalog No. p-200087339 Shop All Labconco Corporation Products
Click to view available options
Includes:
Cabinet, Accessory Package, Scope-Ready
Cabinet, Accessory Package, Scope-Ready, Base Stand
Cabinet, Accessory Package, Scope-Ready, Temp-Zone
Cabinet, Accessory Package, Scope-Ready, Temp-Zone, Base Stand
Cabinet, Accessory Package, Temp-Zone
Cabinet, Accessory Package, Temp-Zone, Base Stand
Plug Type:
North America (115 V, 15 A)
North America (230 V)

Catalog No. 01-812-463

Supplier: Labconco™ 342491002

Only null left
Add to Cart
Add to Cart
Request a Quote

Cell Logics are designed to meet the specific needs of cell research. Models feature a modified sash to accommodate a microscope and/or a temperature-controlled work area on the work surface to maintain cell viability. All models ensure the highest standard of safety and performance.

Performance Features:

  • Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection.
  • Cabinet interior provides ISO Class 5 (Grade A) or better working conditions.
  • Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load.
  • Optimized airflow minimizes energy use and noise output to industry-leading levels.
  • Logic OS with 5 in. line-of-sight touchscreen display.
    • Access cabinet information such as filter life, alerts and alarms, data logs.
    • Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet.
  • Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work.
  • Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work.
  • Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
  • Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
  • Full, six-year warranty on all Class II biosafety cabinets.

Optional Configurations:

  • Scope-Ready™ Package consisting of 7.0 in. wide x 10.5 in. high (18 x 27 cm) sash cutout and Pure-Vu™ Seal with powder-coated steel collar.
    • Stand-Still™ Isolation Platform is required (sold separately).
  • Temp-Zone™ Package consisting of temperature-controlled work area, 10.5 in. wide x 12.0 in. deep (27 x 30 cm) located on the left side of the work surface.
    • Water circulator is required (not included).
  • Telescoping base stand with shelf and leveling feet.

Specifications

Description 4 ft. Cell Logic Class II, Type A2 Biosafety Cabinet
Certifications/Compliance ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2
Chamber Material Stainless Steel (304)
Width (Metric) Interior 123.2 cm
Product Type Class II, Type A2 Bio Safety Cabinet
Model Cell Logic
Noise Level 60 dBA
Outlets 2 Duplex
Plug Type North America (115 V, 15 A)
Power Consumption 195 W
Receptacles 2 Double, Left and Right Side
Height (English) 61.9 in.
Sash Opening (Metric) 22.9 cm
Width (English) Exterior 54.5 in.
Volume (Metric) Exhaust 720 m³/hr.
Display 5 in. touchscreen
Width (Metric) Exterior 138.2 cm
Includes Cabinet, Accessory Package, Scope-Ready, Temp-Zone
Energy Consumption (Reduced Flow Mode) 37 W
Lighting LED, UV
Height (Metric) 157.2 cm
Sash Opening (English) 9 in.
Volume 424 CFM
Width (English) Interior 48.5 in.
Height (English) Interior 25.7 in.
Height (English) Max. Opening 22.5 in.
Height (English) Viewing 27 in.
Height (Metric) Interior 65.3 cm
Height (Metric) Max. Opening 57.2 cm
Height (Metric) Viewing 69 cm
Depth (English) 32 in.
Depth (Metric) 81.3 cm
Voltage 115 V
Energy Consumption (Operating Set Point) 195 W
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.