Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ 4 ft. Purifier Axiom Class II Type C1 Biosafety Cabinet with 10 in. Sash Opening
4 ft. Purifier Axiom Class II Type C1 Biosafety Cabinet with 10 in. Sash Opening is an unique, flexible BSC that can be operated as Type A or B and is the only Type C1 in the world.
Supplier: Labconco™ 304411121
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
- Requires supporting base if included stand option is not selected
- Intrinsically-safe negative pressure design includes:
- Dual Electronically Commutated Motors (ECM)
- Constant Airflow Profile™ (CAP) Technology airflow monitoring system
- Active Protection Protocol operates the cabinet's exhaust blowers for up to five minutes preventing the flow of air into the laboratory in the event of exhaust system failure
- BSC required Air-Wave™ Entry System at access opening to enhance containment
- Contain-Air™ Negative Pressure Channel at top of sash to enhance containment
- Two programmable working sash heights (8 in. and 10 in.)
- 10° angled, fully-closing, 1/4 in. tempered safety glass sash with counterbalanced, anti-racking, easy-to-lift mechanism
- Curved waterfall design stainless steel inlet grille with Reserve-Air™ Secondary Airflow Slots
- 22.6 in. (58 cm) maximum sash opening height and 27.0 in. (69 cm) viewing height Chem-Zone™ work surface with dedicated direct exhaust for use with hazardous vapors or radionuclides
- Leak-tight interior with removable, type 304 stainless steel 3-piece work surface (dished Chem-Zone in center and two flat sides) with lift out knobs and clearly delineated working area
- Nominal inflow velocity of 105 feet per minute (fpm) (0.53 m/sec)
- Nominal downflow velocity of 65 fpm (0.33 m/sec) for 4 ft. models and 55 fpm (0.28 m/sec) for 6 ft. models
- Touchpad on right-hand side post controls blower, light(s), timer, electrical receptacles, audible alarm mute and menu selection
- Omni-Flex™ two mode design (Type A and B mode)
- Low static pressure and volumetric rate exhaust requirements when in Type B mode
- Night-Smart™ System for idling the blower when the sash is fully closed (while in Type A mode only)
- MyLogic™ Operating System and interior-mounted, line-of-sight, full color, easy-to-understand LCD information center with:
- Smart-Start™ System for start up and shut down operations
- Filter Life Remaining bar graph for each HEPA filter
- Status line for alarm conditions and alerts to warn when filter life diminishes to 20% and 0%
- Bright, 90-150 ft. candle, glare-free LED lighting located outside the contaminated work area
- Interval/elapsed timer for experiment monitoring, LED light or UV light control (on models with UV light)
- Electronic security lock (optional activation) that requires access code to operate the cabinet
Specifications
Class II Type C1 Biosafety Cabinet | |
Stainless Steel | |
2 | |
2 | |
Full-color LCD | |
HEPA | |
Powder Coated Steel | |
LED | |
One | |
64.2 in. | |
32.7 in. | |
636 lb. | |
100 V, 50/60 Hz, 12 A |
NSF/ANSI Standard 49, UL Standard 61010-1, CAN/CSA 22.2 No. 61010-1, CE Conformity Marking (230 V models), ISO 5 conditions, ISO 14644-1 and 2, ADA-compliant operation | |
Axiom | |
Japan | |
54.3 in. | |
0.9999 | |
137.9 cm | |
Base Stand, Accessory Pk. | |
10 in. | |
5 Years | |
163.1 cm | |
83.1 cm | |
378.75 kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction