Learn More
Labconco™ 4 ft. Purifier Logic+ Class II Type A2 Biosafety Cabinet with 12 in. Sash Opening
4 ft. Purifier Logic+ Class II Type A2 Biosafety Cabinet with 12 in. Sash Opening creates a particulate-free environment for small animal research activities such as cage changing, weighing, anesthesia delivery and surgical procedures.
$12270.00 - $13635.00
Specifications
Certifications/Compliance NSF/ANSI Standard 49, UL Standard 61010-1, CAN/CSA 22.2 No. 61010-1, CE Conformity Marking (230 V models), ISO 5 conditions, ISO 14644-1 and 2, ADA-compliant operation
Item Description Class II Type A2 Biosafety Cabinet
View More Specs
Products 20
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Price Quantity & Availability  
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Price Quantity & Availability  
30-242-0021
View Documents
Labconco™
302420021
Japan 100 V, 50/60 Hz, 12 A Biosafety Cabinet only
Each for $12,270.00
Add to cart
30-242-0031
View Documents
Labconco™
302420031
Schuko 230 V, 50/60 Hz, 6 A Biosafety Cabinet only
Each for $12,270.00
Add to cart
30-242-0041
View Documents
Labconco™
302420041
China/Australia 230 V, 50/60 Hz, 6 A Biosafety Cabinet only
Each for $12,270.00
Add to cart
30-242-0051
View Documents
Labconco™
302420051
British (UK) 230 V, 50/60 Hz, 6 A Biosafety Cabinet only
Each for $12,270.00
Add to cart
30-242-0071
View Documents
Labconco™
302420071
India 230 V, 50/60 Hz, 6 A Biosafety Cabinet only
Each for $12,270.00
Add to cart
30-242-0121
View Documents
Labconco™
302420121
Japan 100 V, 50/60 Hz, 12 A Base Stand
Each for $12,970.00
Add to cart
30-242-0131
View Documents
Labconco™
302420131
Schuko 230 V, 50/60 Hz, 6 A Base Stand
Each for $12,970.00
Add to cart
30-242-0141
View Documents
Labconco™
302420141
China/Australia 230 V, 50/60 Hz, 6 A Base Stand
Each for $12,970.00
Add to cart
30-242-0151
View Documents
Labconco™
302420151
British (UK) 230 V, 50/60 Hz, 6 A Base Stand
Each for $12,970.00
Add to cart
30-242-0171
View Documents
Labconco™
302420171
India 230 V, 50/60 Hz, 6 A Base Stand
Each for $12,970.00
Add to cart
30-242-1021
View Documents
Labconco™
302421021
Japan 100 V, 50/60 Hz, 12 A Accessory Pk.
Each for $12,945.00
Add to cart
30-242-1031
View Documents
Labconco™
302421031
Schuko 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $12,945.00
Add to cart
30-242-1041
View Documents
Labconco™
302421041
China/Australia 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $12,945.00
Add to cart
30-242-1051
View Documents
Labconco™
302421051
British (UK) 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $12,945.00
Add to cart
30-242-1071
View Documents
Labconco™
302421071
India 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $12,945.00
Add to cart
30-242-1121
View Documents
Labconco™
302421121
Japan 100 V, 50/60 Hz, 12 A Base Stand and Accessory Pk.
Each for $13,635.00
Add to cart
30-242-1131
View Documents
Labconco™
302421131
Schuko 230 V, 50/60 Hz, 6 A Base Stand and Accessory Pk.
Each for $13,635.00
Add to cart
30-242-1141
View Documents
Labconco™
302421141
China/Australia 230 V, 50/60 Hz, 6 A Base Stand and Accessory Pk.
Each for $13,635.00
Add to cart
30-242-1151
View Documents
Labconco™
302421151
British (UK) 230 V, 50/60 Hz, 6 A Base Stand and Accessory Pk.
Each for $13,635.00
Add to cart
30-242-1171
View Documents
Labconco™
302421171
India 230 V, 50/60 Hz, 6 A Base Stand and Accessory Pk.
Each for $13,635.00
Add to cart
Description
Description
  • Purifier Logic+ Class II A2 Biosafety Cabinets provide personnel, product and environmental protection and are suitable for work in all biosafety levels
  • Designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection
  • Some models have base stands and/or an Accessory Package (UV light, service fixture(s) and Vacu-Pass™ Cord and Cable Portal) included
  • Inclination™ Technology Suite that includes MyLogic™ operating system with eye-level color display, 10° angled sash, patented curved air grille with built in armrest, plus fully accessible controls, outlets, and fixtures
  • MyLogic is easy to use, displaying the BSC's status with clear messaging and graphics that notify users of filter life, alerts, and alarms
  • Energy-efficient ECM blower powered by Labconco’s exclusive Constant Airflow Profile™ (CAP) technology
Specifications
Specifications
NSF/ANSI Standard 49, UL Standard 61010-1, CAN/CSA 22.2 No. 61010-1, CE Conformity Marking (230 V models), ISO 5 conditions, ISO 14644-1 and 2, ADA-compliant operation
Class II Type A2 Biosafety Cabinet