Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Guanidinobenzoic acid hydrochloride, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
42823-46-1
C8H10ClN3O2
215.637
MFCD00040587
YETFLAUJROGBMC-UHFFFAOYSA-N
4-guanidinobenzoic acid hydrochloride,n-4-carboxyphenyl guanidine hydrochloride,4-guanidinobenzoic acid hcl,4-guanidino benzoic acid hydrochloride,4-amidinoamino benzoic acid, chloride,pubchem14981,pubchem14993,acmc-1an3m,ksc494a0j,4-guanidino-benzoic acid-hcl salt
3084875
4-(diaminomethylideneamino)benzoic acid;hydrochloride
C1=CC(=CC=C1C(=O)O)N=C(N)N.Cl
42823-46-1
C8H10ClN3O2
215.637
MFCD00040587
YETFLAUJROGBMC-UHFFFAOYSA-N
4-guanidinobenzoic acid hydrochloride,n-4-carboxyphenyl guanidine hydrochloride,4-guanidinobenzoic acid hcl,4-guanidino benzoic acid hydrochloride,4-amidinoamino benzoic acid, chloride,pubchem14981,pubchem14993,acmc-1an3m,ksc494a0j,4-guanidino-benzoic acid-hcl salt
3084875
4-(diaminomethylideneamino)benzoic acid;hydrochloride
C1=CC(=CC=C1C(=O)O)N=C(N)N.Cl
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Color White to Beige
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula NH2C(NH)NHC6H4COOH
Formula Weight 215.64
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material 4-Guanidinobenzoic acid hydrochloride, 95%
Show More Show Less

EINECSNumber : 255-956-3