Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Heptanone, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
500mL
2.5L
Packaging:
Glass bottle
123-19-3
C7H14O
114.19
MFCD00009403
HCFAJYNVAYBARA-UHFFFAOYSA-N
4-heptanone,dipropyl ketone,propyl ketone,butyrone,di-n-propyl ketone,4-heptanone natural,4-oxoheptane,unii-9bn582jq61,fema no. 2546,dipropylketon
31246
heptan-4-one
CCCC(=O)CCC
123-19-3
C7H14O
114.19
MFCD00009403
HCFAJYNVAYBARA-UHFFFAOYSA-N
4-heptanone,dipropyl ketone,propyl ketone,butyrone,di-n-propyl ketone,4-heptanone natural,4-oxoheptane,unii-9bn582jq61,fema no. 2546,dipropylketon
31246
heptan-4-one
CCCC(=O)CCC
Viscosity 0.73 mPa.s (20°C)
Boiling Point 145.0°C
Refractive Index 1.4060 to 1.4080
Quantity 5mL
Packaging Glass bottle
Specific Gravity 0.81
Infrared Spectrum Authentic
Flash Point 48°C
Color Colorless to Yellow
Density 0.8100g/mL
Melting Point -33.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CH2CH2COCH2CH2CH3
Beilstein 01,699
Merck Index 15,3386
Solubility Information Solubility in water: 4.6g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 114.19
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Heptanone, 98%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-608-9

RTECSNumber : MJ5600000

TSCA : TSCA