Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexen-1-ol, 97%, predominantly trans, ACROS Organics™

Click to view available options
Quantity:
5g
25g
928-92-7
C6H12O
100.16
VTIODUHBZHNXFP-NSCUHMNNSA-N
4-hexen-1-ol,e-4-hexen-1-ol,e-hex-4-en-1-ol,trans-4-hexen-1-ol,4-hexen-1-ol, 4e,4-hexen-1-ol, e,trans-4-hexenol,unii-zhb2532c2h,4-hexen-1-ol, predominantly trans
641248
(E)-hex-4-en-1-ol
CC=CCCCO
928-92-7
C6H12O
100.16
VTIODUHBZHNXFP-NSCUHMNNSA-N
4-hexen-1-ol,e-4-hexen-1-ol,e-hex-4-en-1-ol,trans-4-hexen-1-ol,4-hexen-1-ol, 4e,4-hexen-1-ol, e,trans-4-hexenol,unii-zhb2532c2h,4-hexen-1-ol, predominantly trans
641248
(E)-hex-4-en-1-ol
CC=CCCCO
Infrared Spectrum Authentic
Flash Point 61°C
Specific Gravity 0.85
Color Colorless
Boiling Point 159.0°C to 160.0°C
Packaging Glass bottle
Quantity 5g
Density 0.8500g/mL
Refractive Index 1.4394
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3CH=CH(CH2)3OH
Beilstein 01,III,1929
Formula Weight 100.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Hexen-1-ol
Show More Show Less

GHS H Statement:

GHS P Statement
Wear eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 213-188-6