Alfa Aesar™ 4-Hexen-3-one, 98%, trans-isomer >95%

Manufacturer: Alfa Aesar™ H2744518

Quantity 50g

 View more versions of this product

Catalog No. AAH2744518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2497-21-4
    Molecular Formula C6H10O
    Formula Weight 98.15
    MDL Number MFCD00010631
    UN Number UN1224
    Chemical Name or Material 4-Hexen-3-one

    Specifications

    Formula Weight 98.15
    TSCA TSCA
    UN Number UN1224
    Odor Characteristic
    Boiling Point 135° to 137°C
    Quantity 50g
    Molecular Formula C6H10O
    MDL Number MFCD00010631
    CAS 2497-21-4
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H302
    Flammable liquid and vapor.
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage −30° to −10°C
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
    RTECS Number MP8659900
    Flash Point 34°C (93°F)
    Chemical Name or Material 4-Hexen-3-one
    Assay 98%, trans-isomer >95%

    GHS H Statement
    H226-H302
    Flammable liquid and vapor.
    Harmful if swallowed.

    −30° to −10°C

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit