Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydrazinobenzenesulfonic acid, hemihydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
100g
Packaging:
Glass bottle
854689-07-9,98-71-5
C6H8N2O31/2H2O
197.21
MFCD00480958
OSOMMFVBTYFMBI-UHFFFAOYSA-N
4-hydrazinylbenzenesulfonic acid hydrate,4-hydrazinobenzenesulfonic acid hydrate,phenylhydrazine-4-sulfonic acid hemihydrate,benzenesulfonic acid, 4-hydrazino-, monohydrate 9ci
2849446
4-hydrazinylbenzenesulfonic acid;hydrate
C1=CC(=CC=C1NN)S(=O)(=O)O.O
854689-07-9,98-71-5
C6H8N2O31/2H2O
197.21
MFCD00480958
OSOMMFVBTYFMBI-UHFFFAOYSA-N
4-hydrazinylbenzenesulfonic acid hydrate,4-hydrazinobenzenesulfonic acid hydrate,phenylhydrazine-4-sulfonic acid hemihydrate,benzenesulfonic acid, 4-hydrazino-, monohydrate 9ci
2849446
4-hydrazinylbenzenesulfonic acid;hydrate
C1=CC(=CC=C1NN)S(=O)(=O)O.O
Quantity 50g
Packaging Glass bottle
Infrared Spectrum Authentic
Loss on Drying 4% to 5%
Melting Point 285.0°C
Color Orange to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 14, 4773
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in alcohol
Formula Weight 197.21
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4-Hydrazinobenzenesulfonic acid, hemihydrate, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-694-2

RTECSNumber : DB6900000

TSCA : TSCA