Learn More
Medchemexpress LLC 4-hydrazinobenzoic acid | 619-67-0 | MFCD00007581 | 100.0% | 152.15 g/mol | C7H8N2O2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01738250G
Encompass_Preferred
4-hydrazinobenzoic acid is an organic biochemical reagent used as a building block in organic synthesis and as an analytical standard in life-science research. It is supplied as a yellow-brown solid and is characterized by CAS 619-67-0 and molecular formula C7H8N2O2.
  • High purity suitable for analytical and synthetic applications.
  • Solid form, yellow to brown, easy to handle and weigh.
  • Stable when stored under recommended conditions for long-term use.
  • Supplied with certificate of analysis and safety data sheet for documentation.
  • Commonly used as a building block in organic synthesis and as an analytical standard.
Catalog No. 50-003-01189