Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Hydroxy-2,4-dimethoxybenzophenone, 98%

Click to view available options
Quantity:
5g
25g
100g
41351-30-8
C15H14O4
258.273
MFCD00137850
QEHRETCJMLQPCR-UHFFFAOYSA-N
2,4-dimethoxy-4'-hydroxybenzophenone,2,4-dimethoxyphenyl 4-hydroxyphenyl methanone,4-2,4-dimethoxybenzoyl phenol,4'-hydroxy-2,4-dimethoxybenzophenone,2,4-dimethoxy-4-hydroxybenzophenone,2,4-dimethoxyphenyl-4-hydroxyphenyl methanone,methanone, 2,4-dimethoxyphenyl 4-hydroxyphenyl,2,4-dimethoxyphenyl 4-hydroxyphenyl ketone,pubchem7483,acmc-209jk0
688628
(2,4-dimethoxyphenyl)-(4-hydroxyphenyl)methanone
COC1=CC(=C(C=C1)C(=O)C2=CC=C(C=C2)O)OC
41351-30-8
C15H14O4
258.273
MFCD00137850
QEHRETCJMLQPCR-UHFFFAOYSA-N
2,4-dimethoxy-4'-hydroxybenzophenone,2,4-dimethoxyphenyl 4-hydroxyphenyl methanone,4-2,4-dimethoxybenzoyl phenol,4'-hydroxy-2,4-dimethoxybenzophenone,2,4-dimethoxy-4-hydroxybenzophenone,2,4-dimethoxyphenyl-4-hydroxyphenyl methanone,methanone, 2,4-dimethoxyphenyl 4-hydroxyphenyl,2,4-dimethoxyphenyl 4-hydroxyphenyl ketone,pubchem7483,acmc-209jk0
688628
(2,4-dimethoxyphenyl)-(4-hydroxyphenyl)methanone
COC1=CC(=C(C=C1)C(=O)C2=CC=C(C=C2)O)OC
Melting Point 138°C to 142°C
Color White
Quantity 5g
Beilstein 2508320
Formula Weight 258.27
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4'-Hydroxy-2,4-dimethoxybenzophenone

storageNote1 : 2° to 8°C