Learn More
4-Hydroxy-2-hexenal Mouse anti-Chemical, Clone: 5C11.1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527576
Catalog No. PIMA527576
Add to cart
Description
Description
4-Hydroxy-2-hexenal is a lipid peroxidation product derived from oxidized omega-3 fatty acids such as DHA (decosahexanoic acid).
Specifications
Specifications
4-Hydroxy-2-hexenal
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
4-Hydroxy-2-hexenal (4-HHE), 4-Hydroxy-2-hexenal, 4-HHE, MG, MG-modified protein, 4-Hydroxy-2-hexenal-modified, 4-Hydroxy-2-hexenal, 4-hydroxy Hexenal, HHE, 4-HHE
Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH)
Protein G
RUO
Antibody
Monoclonal
ELISA, Western Blot
5C11.1
Unconjugated
Liquid
Mouse
IgG1
100 μg
-20°C
Primary
Chemical