Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Hydroxynonenal Mouse anti-Nonspecies specific, HRP, Clone: 12F7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259353H
Description
4-Hydroxynonenal Monoclonal specifically detects 4-Hydroxynonenal in Non-species specific samples. It is validated for Western Blot, ELISA.Specifications
4-Hydroxynonenal | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
Protein G purified | |
Primary | |
Nonspecies specific | |
Purified |
Western Blot, ELISA | |
12F7 | |
Western Blot, ELISA | |
(2E)-4-Hydroxy-2-nonenal, 2-Nonenal, 4-hydroxy-, (2E,4R)-, 4 HNE, 4-hydroxy-trans-2-nonenal | |
Synthetic 4-Hydroxynonenal modified Keyhole Limpet Hemocyanin (KLH) | |
100 μL | |
Specific for 4-Hydroxynonenal (4-HNE) modified proteins. Does not detect free 4-Hydroxynonenal. Does not cross-react with 4-Hydroxy-2-hexenal, Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins. | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title