Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-2-hexenal Mouse anti-Nonspecies specific, PE/Cy5.5, Clone: 6F10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259352PECY55

Catalog No. N259352PEC5


Add to cart

Description

Description

4-Hydroxy-2-hexenal Monoclonal antibody specifically detects 4-Hydroxy-2-hexenal in Nonspecies specific samples. It is validated for Flow Cytometry.
Specifications

Specifications

4-Hydroxy-2-hexenal
6F10
Flow Cytometry
Conjugated, Purified
Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Nonspecies specific
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
Mouse
IgG2b
0.1 mL
Primary
Specific for 4-Hydroxy-2-hexenal (4-HHE) and 4-Hydroxynonenal (4-HNE) modified proteins. Does not detect free 4-Hydroxy-2-hexenal (4-HHE) or 4-Hydroxynonenal (4-HNE). Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit