Learn More
4-Hydroxy-2-hexenal Mouse, ATTO 390, Clone: 6F10, Abnova
Mouse Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB19269
Description
4-Hydroxy-2-hexenal Monoclonal antibody specifically detects 4-Hydroxy-2-hexenal in samples. It is validated for WB, ELISA, ICC, IF, Flow Cyt.Specifications
4-Hydroxy-2-hexenal | |
6F10 | |
Liquid | |
Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH) | |
100 μg | |
Primary |
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
ATTO 390 | |
Mouse | |
IgG2b | |
Store at -20°C | |
Monoclonal |