Learn More
4-Hydroxy-2-hexenal Mouse, ATTO 655, Clone: 6F10, Abnova™
Mouse Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB19274
Catalog No. 89-338-778
Add to cart
Description
Description
4-Hydroxy-2-hexenal Monoclonal antibody specifically detects 4-Hydroxy-2-hexenal in samples. It is validated for WB, ELISA, ICC, IF, Flow Cyt.
Specifications
Specifications
4-Hydroxy-2-hexenal
6F10
Liquid
Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH)
100 μg
Primary
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
ATTO 655
Mouse
IgG2b
Store at -20°C
Monoclonal