Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Hydroxy-2'-methylacetophenone, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
875-59-2
C9H10O2
150.18
MFCD00002303
IAMNVCJECQWBLZ-UHFFFAOYSA-N
4'-hydroxy-2'-methylacetophenone,1-4-hydroxy-2-methylphenyl ethanone,4-hydroxy-2-methylacetophenone,2-methyl-4-hydroxyacetophenone,ethanone, 1-4-hydroxy-2-methylphenyl,3-methyl-4-acetylphenol,1-4-hydroxy-2-methylphenyl ethan-1-one,1-acetyl-4-hydroxy-2-methylbenzene,ethanone,1-4-hydroxy-2-methylphenyl,2'-methyl-4'-hydroxyacetophenone
70133
CHEBI:87314
1-(4-hydroxy-2-methylphenyl)ethanone
CC1=C(C=CC(=C1)O)C(=O)C
875-59-2
C9H10O2
150.18
MFCD00002303
IAMNVCJECQWBLZ-UHFFFAOYSA-N
4'-hydroxy-2'-methylacetophenone,1-4-hydroxy-2-methylphenyl ethanone,4-hydroxy-2-methylacetophenone,2-methyl-4-hydroxyacetophenone,ethanone, 1-4-hydroxy-2-methylphenyl,3-methyl-4-acetylphenol,1-4-hydroxy-2-methylphenyl ethan-1-one,1-acetyl-4-hydroxy-2-methylbenzene,ethanone,1-4-hydroxy-2-methylphenyl,2'-methyl-4'-hydroxyacetophenone
70133
CHEBI:87314
1-(4-hydroxy-2-methylphenyl)ethanone
CC1=C(C=CC(=C1)O)C(=O)C
Boiling Point 313°C
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 127°C to 132°C
Color Tan to White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HOC6H3(CH3)COCH3
Beilstein 08, 111
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 150.18
Physical Form Powder or Crystals
Percent Purity ≥96.0%
Chemical Name or Material 4'-Hydroxy-2'-methylacetophenone
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning