Alfa Aesar™ 4-[Hydroxy(2-thiazolyl)methyl]benzeneboronic acid pinacol ester, 97%

Manufacturer: Alfa Aesar™ H2879806

Quantity 5g

 View more versions of this product

Catalog No. AAH2879806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1073371-73-9
    Molecular Formula C16H20BNO3S
    Formula Weight 317.21
    MDL Number MFCD09953504
    Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-alpha-(2-thiazolyl)benzyl alcohol; [alpha-(2-Thiazolyl)-4-(hydroxymethyl)]benzeneboronic acid pinacol ester
    Chemical Name or Material 4-[Hydroxy(2-thiazolyl)methyl]benzeneboronic acid pinacol ester

    Specifications

    Formula Weight 317.21
    Melting Point 147° to 150°C
    Quantity 5g
    Molecular Formula C16H20BNO3S
    MDL Number MFCD09953504
    CAS 1073371-73-9
    Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-alpha-(2-thiazolyl)benzyl alcohol; [alpha-(2-Thiazolyl)-4-(hydroxymethyl)]benzeneboronic acid pinacol ester
    Chemical Name or Material 4-[Hydroxy(2-thiazolyl)methyl]benzeneboronic acid pinacol ester
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit