Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Hydroxy-2-(trifluoromethyl)pyridine, 97%

Click to view available options
Quantity:
250mg
1g
5g
170886-13-2
C6H4F3NO
163.099
MFCD01571202
LKHCWMQMRXAPHX-UHFFFAOYSA-N
2-trifluoromethyl pyridin-4-ol,4-hydroxy-2-trifluoromethyl pyridine,4-hydroxy-2-trifluoromethylpyridine,2-trifluoromethyl-4-hydroxypyridine,2-trifluoromethyl-4-pyridinol,4-pyridinol, 2-trifluoromethyl,2-trifluoromethyl-1h-pyridin-4-one,2-trifluoromethylpyridin-4-ol,2-trifluoromethyl-1,4-dihydropyridin-4-one,pubchem17703
10953849
2-(trifluoromethyl)-1H-pyridin-4-one
C1=CNC(=CC1=O)C(F)(F)F
170886-13-2
C6H4F3NO
163.099
MFCD01571202
LKHCWMQMRXAPHX-UHFFFAOYSA-N
2-trifluoromethyl pyridin-4-ol,4-hydroxy-2-trifluoromethyl pyridine,4-hydroxy-2-trifluoromethylpyridine,2-trifluoromethyl-4-hydroxypyridine,2-trifluoromethyl-4-pyridinol,4-pyridinol, 2-trifluoromethyl,2-trifluoromethyl-1h-pyridin-4-one,2-trifluoromethylpyridin-4-ol,2-trifluoromethyl-1,4-dihydropyridin-4-one,pubchem17703
10953849
2-(trifluoromethyl)-1H-pyridin-4-one
C1=CNC(=CC1=O)C(F)(F)F
Color White
Quantity 250mg
Formula Weight 163.1
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-2-(trifluoromethyl)pyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning