Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Hydroxy-3,5-dimethoxycinnamic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
530-59-6
C11H12O5
224.212
MFCD00004401
PCMORTLOPMLEFB-ONEGZZNKSA-N
sinapic acid,sinapinic acid,3,5-dimethoxy-4-hydroxycinnamic acid,sinapinate,sinapate,trans-sinapic acid,4-hydroxy-3,5-dimethoxycinnamic acid,synapoic acid,2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid,unii-p0i60993ec
637775
CHEBI:15714
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
COC1=CC(=CC(=C1O)OC)C=CC(=O)O
530-59-6
C11H12O5
224.212
MFCD00004401
PCMORTLOPMLEFB-ONEGZZNKSA-N
sinapic acid,sinapinic acid,3,5-dimethoxy-4-hydroxycinnamic acid,sinapinate,sinapate,trans-sinapic acid,4-hydroxy-3,5-dimethoxycinnamic acid,synapoic acid,2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid,unii-p0i60993ec
637775
CHEBI:15714
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
COC1=CC(=CC(=C1O)OC)C=CC(=O)O
Applications
4-Hydroxy-3,5-dimethoxycinnamic acid is a commonly used matrix in MALDI mass spectrometry. It is a useful matrix for a wide variety of peptides and proteins. It serves well as a matrix for MALDI due to its ability to absorb laser radiation and to also donate protons (H+) to the analyte of interest.

Solubility
Soluble in methanol, ethanol and dimethyl sulfoxide.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Quantity 5g
Melting Point ~195°C (decomposition)
Beilstein 2130006
Solubility Information Soluble in methanol,ethanol and dimethyl sulfoxide.
Formula Weight 224.21
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3,5-dimethoxycinnamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-487-3

TSCA : No

Recommended Storage : Ambient temperatures