Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3,5-dimethylbenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
4919-37-3
C9H10O3
166.18
MFCD00016536
OMNHTTWQSSUZHO-UHFFFAOYSA-N
3,5-dimethyl-4-hydroxybenzoic acid,benzoic acid, 4-hydroxy-3,5-dimethyl,pubchem16247,acmc-209kf7,ksc496e2b,omnhttwqssuzho-uhfffaoysa,3.5-dimethyl-4-hydroxybenzoic acid,3,5-dimethyl-4-hydroxy-benzoic acid,4-hydroxy-3,5-dimethyl-benzoic acid
138387
4-hydroxy-3,5-dimethylbenzoic acid
CC1=CC(=CC(=C1O)C)C(=O)O
4919-37-3
C9H10O3
166.18
MFCD00016536
OMNHTTWQSSUZHO-UHFFFAOYSA-N
3,5-dimethyl-4-hydroxybenzoic acid,benzoic acid, 4-hydroxy-3,5-dimethyl,pubchem16247,acmc-209kf7,ksc496e2b,omnhttwqssuzho-uhfffaoysa,3.5-dimethyl-4-hydroxybenzoic acid,3,5-dimethyl-4-hydroxy-benzoic acid,4-hydroxy-3,5-dimethyl-benzoic acid
138387
4-hydroxy-3,5-dimethylbenzoic acid
CC1=CC(=CC(=C1O)C)C(=O)O
Infrared Spectrum Authentic
Melting Point 221.0°C to 226.0°C
Color Beige or Pink to White
Packaging Glass bottle
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 166.18
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3, 5-dimethylbenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning