Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Hydroxy-3-methoxybenzyl alcohol, 98+%

Manufacturer:  Alfa Aesar™ A1516018

Quantity 50g

 View more versions of this product

Catalog No. AAA1516018


  Check Availability
Add to cart

Specifications

Description

Applications
Used as a flavoring agent. Other possible uses, vanilla, coconut, cream and other dairy nuances, coumarin.

Solubility
Soluble in 95% ethanol (5 %), water, and oils.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

498-00-0
154.17
1910044
4-Hydroxy-3-methoxybenzyl alcohol
C8H10O3
MFCD00004659
Vanillyl alcohol

Specifications

113° to 115°C
498-00-0
154.17
1910044
4-Hydroxy-3-methoxybenzyl alcohol
50g
C8H10O3
MFCD00004659
Vanillyl alcohol
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit