Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-methoxybenzyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Plastic bottle
498-00-0
C8H10O3
154.16
MFCD00004659
ZENOXNGFMSCLLL-UHFFFAOYSA-N
vanillyl alcohol,4-hydroxy-3-methoxybenzyl alcohol,4-hydroxymethyl-2-methoxyphenol,vanillin alcohol,vanillic alcohol,4-hydroxy-3-methoxybenzenemethanol,benzenemethanol, 4-hydroxy-3-methoxy,vanillol,4-hydroxy-3-methoxybenzylalcohol,4-hydroxy-3-methoxyphenylmethanol
62348
CHEBI:18353
4-(hydroxymethyl)-2-methoxyphenol
COC1=C(C=CC(=C1)CO)O
498-00-0
C8H10O3
154.16
MFCD00004659
ZENOXNGFMSCLLL-UHFFFAOYSA-N
vanillyl alcohol,4-hydroxy-3-methoxybenzyl alcohol,4-hydroxymethyl-2-methoxyphenol,vanillin alcohol,vanillic alcohol,4-hydroxy-3-methoxybenzenemethanol,benzenemethanol, 4-hydroxy-3-methoxy,vanillol,4-hydroxy-3-methoxybenzylalcohol,4-hydroxy-3-methoxyphenylmethanol
62348
CHEBI:18353
4-(hydroxymethyl)-2-methoxyphenol
COC1=C(C=CC(=C1)CO)O
Packaging Plastic bottle
Infrared Spectrum Authentic
Melting Point 112.0°C to 115.0°C
Color White to Beige
Quantity 50g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula HOC6H3(OCH3)CH2OH
Beilstein 06,IV,7381
Formula Weight 154.16
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-Hydroxy-3-methoxybenzyl alcohol, 99%
Show More Show Less

EINECSNumber : 207-852-4