Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Hydroxy-3-methyl-2-butanone, tech 85%

Manufacturer:  Alfa Aesar™ H2848306

Quantity 5g

 View more versions of this product

Catalog No. AAH2848306


  Check Availability
Add to cart

Specifications

Description

Applications
Protonated diacetone alcohol and protonated 4- hydroxy-3-methyl-2-butanone undergo “vinylation” and colli- sion-induced dehydration as the only primary reactions, the product of the former reaction consisting of up to 40% of the total product ion distribution.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

3393-64-4
102.13
4-Hydroxy-3-methyl-2-butanone
C5H10O2
MFCD00004739
Technical Grade

Specifications

5g
C5H10O2
MFCD00004739
85%
3393-64-4
102.13
4-Hydroxy-3-methyl-2-butanone
Technical Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit