Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Hydroxy-3'-methylacetophenone, 98%

Click to view available options
Quantity:
1g
5g
25g
876-02-8
C9H10O2
150.177
MFCD00002231
LXBHHIZIQVZGFN-UHFFFAOYSA-N
4'-hydroxy-3'-methylacetophenone,4-hydroxy-3-methylacetophenone,1-4-hydroxy-3-methylphenyl ethanone,1-4-hydroxy-3-methylphenyl ethan-1-one,4-acetyl-2-methylphenol,ethanone, 1-4-hydroxy-3-methylphenyl,4-hydroxy-3-methyl acetophenone,1-4-hydroxy-3-methyl-phenyl-ethanone,1-acetyl-4-hydroxy-3-methylbenzene
70135
1-(4-hydroxy-3-methylphenyl)ethanone
CC1=C(C=CC(=C1)C(=O)C)O
876-02-8
C9H10O2
150.177
MFCD00002231
LXBHHIZIQVZGFN-UHFFFAOYSA-N
4'-hydroxy-3'-methylacetophenone,4-hydroxy-3-methylacetophenone,1-4-hydroxy-3-methylphenyl ethanone,1-4-hydroxy-3-methylphenyl ethan-1-one,4-acetyl-2-methylphenol,ethanone, 1-4-hydroxy-3-methylphenyl,4-hydroxy-3-methyl acetophenone,1-4-hydroxy-3-methyl-phenyl-ethanone,1-acetyl-4-hydroxy-3-methylbenzene
70135
1-(4-hydroxy-3-methylphenyl)ethanone
CC1=C(C=CC(=C1)C(=O)C)O
Quantity 25g
Melting Point 107°C to 109°C
Boiling Point 175°C (1mmHg)
Beilstein 2041839
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 4'-Hydroxy-3'-methylacetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 212-880-5

TSCA : No

Recommended Storage : Ambient temperatures